Assay Details
| Gene Symbol: PYCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060222.1 | XP_017915711.1 | 2 - 3 | 55 | ||
| XM_018060223.1 | XP_017915712.1 | 3 - 4 | 55 | |||
| GenBank mRNA | JO433359.2 | - | 3 - 4 | 464 | 55 | |
More Information
Set Membership: |
|

PYCR2
Genomic Map