Assay Details
| Gene Symbol: PLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065725.1 | XP_017921214.1 | 10 - 11 | 1630 | 57 | |
| GenBank mRNA | KA343795.1 | - | 4 - 5 | 1145 | 57 | |
More Information
Set Membership: |
|

PLK2
Genomic Map