Assay Details
Gene Symbol: USP49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038729.1 | XP_017894218.1 | 1 - 2 | 1366 | 85 | |
XM_018038730.1 | XP_017894219.1 | 1 - 2 | 1366 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
