Assay Details
| Gene Symbol: NAGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048832.1 | XP_017904321.1 | 4 - 5 | 537 | 68 | |
| XM_018048833.1 | XP_017904322.1 | 4 - 5 | 480 | 68 | ||
More Information
Set Membership: |
|

NAGA
Genomic Map