Assay Details
| Gene Symbol: TECRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681648.2 | XP_005681705.2 | 6 - 7 | 755 | 82 | |
| GenBank mRNA | JO392685.2 | - | 6 - 7 | 440 | 82 | |
More Information
Set Membership: |
|

TECRL
Genomic Map