Assay Details
| Gene Symbol: AURKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688324.3 | XP_005688381.2 | 8 - 9 | 1164 | 62 | |
| GenBank mRNA | JO598705.2 | - | 8 - 9 | 1079 | 62 | |
More Information
Set Membership: |
|

AURKA
Genomic Map