Assay Details
| Gene Symbol: TCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690335.3 | XP_005690392.1 | 7 - 8 | 1111 | 69 | |
| GenBank mRNA | KA333597.1 | - | 2 - 3 | 237 | 69 | |
More Information
Set Membership: |
|

TCN1
Genomic Map