Assay Details
| Gene Symbol: FLVCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690999.3 | XP_005691056.2 | 6 - 7 | 1659 | 65 | |
| GenBank mRNA | JO594756.2 | - | 4 - 5 | 317 | 65 | |
| JT709525.1 | - | 3 - 4 | 652 | 65 | ||
More Information
Set Membership: |
|

FLVCR1
Genomic Map