Assay Details
| Gene Symbol: PRRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692725.2 | XP_005692782.1 | 6 - 7 | 603 | 65 | |
| XM_013971575.2 | XP_013827029.1 | 5 - 6 | 612 | 65 | ||
| GenBank mRNA | JO590890.2 | - | 6 - 7 | 546 | 65 | |
More Information
Set Membership: |
|

PRRG2
Genomic Map