Assay Details
| Gene Symbol: SCIMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693445.3 | XP_005693502.1 | 4 - 5 | 383 | 118 | |
| XM_005693446.3 | XP_005693503.1 | 4 - 5 | 1265 | 118 | ||
| XM_013972056.2 | XP_013827510.1 | 4 - 5 | 383 | 118 | ||
| GenBank mRNA | JO603801.2 | - | 4 - 5 | 375 | 118 | |
More Information
Set Membership: |
|

SCIMP
Genomic Map