Assay Details
| Gene Symbol: LOC102175795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064842.1 | XP_017920331.1 | 2 - 3 | 360 | 56 | |
| XM_018064843.1 | XP_017920332.1 | 2 - 3 | 320 | 56 | ||
| GenBank mRNA | JO556810.2 | - | 2 - 3 | 218 | 56 | |
| JO580522.1 | - | 2 - 3 | 297 | 56 | ||
More Information
Set Membership: |
|

LOC102175795
Genomic Map