Assay Details
| Gene Symbol: CPEB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694540.3 | XP_005694597.1 | 5 - 6 | 2493 | 60 | |
| XM_005694541.3 | XP_005694598.1 | 4 - 5 | 2468 | 60 | ||
| XM_005694543.3 | XP_005694600.1 | 3 - 4 | 2417 | 60 | ||
More Information
Set Membership: |
|

CPEB4
Genomic Map