Assay Details
| Gene Symbol: INKA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695942.3 | XP_005695999.2 | 1 - 2 | 248 | 114 | |
| GenBank mRNA | JO523442.2 | - | 1 - 2 | 327 | 114 | |
| JO608290.2 | - | 1 - 2 | 875 | 114 | ||
| KA324530.1 | - | 1 - 2 | 216 | 114 | ||
More Information
Set Membership: |
|

INKA1
Genomic Map