Assay Details
| Gene Symbol: CD2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697699.2 | XP_005697756.1 | 2 - 3 | 221 | 60 | |
| XM_013974750.2 | XP_013830204.1 | 2 - 3 | 475 | 60 | ||
| XM_013974751.2 | XP_013830205.1 | 2 - 3 | 256 | 60 | ||
More Information
Set Membership: |
|

CD2BP2
Genomic Map