Assay Details
| Gene Symbol: LGALS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042238.1 | XP_017897727.1 | 3 - 4 | 502 | 68 | |
| XM_018042239.1 | XP_017897728.1 | 3 - 4 | 502 | 68 | ||
| GenBank mRNA | JO421129.2 | - | 7 - 8 | 2270 | 68 | |
| JO509639.2 | - | 6 - 7 | 613 | 68 | ||
More Information
Set Membership: |
|

LGALS8
Genomic Map