Assay Details
| Gene Symbol: DRAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699908.3 | XP_005699965.2 | 5 - 6 | 666 | 63 | |
| GenBank mRNA | JO581745.1 | - | 2 - 3 | 349 | 63 | |
| JO631807.2 | - | 2 - 3 | 162 | 63 | ||
More Information
Set Membership: |
|

DRAP1
Genomic Map