Assay Details
| Gene Symbol: PSMD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700085.3 | XP_005700142.1 | 7 - 8 | 651 | 66 | |
| XM_018044421.1 | XP_017899910.1 | 7 - 8 | 652 | 66 | ||
| GenBank mRNA | JO631935.2 | - | 7 - 8 | 975 | 66 | |
| KA335914.1 | - | 7 - 8 | 955 | 66 | ||
More Information
Set Membership: |
|

PSMD13
Genomic Map