Assay Details
| Gene Symbol: NUP62CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700211.3 | XP_005700268.1 | 2 - 3 | 228 | 85 | |
| GenBank mRNA | JO438094.2 | - | 3 - 4 | 259 | 85 | |
More Information
Set Membership: |
|

NUP62CL
Genomic Map