Assay Details
| Gene Symbol: PNPLA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049122.1 | XP_017904611.1 | 3 - 4 | 587 | 63 | |
| XM_018049123.1 | XP_017904612.1 | 3 - 4 | 587 | 63 | ||
| GenBank mRNA | KA334638.1 | - | 3 - 4 | 442 | 63 | |
More Information
Set Membership: |
|

PNPLA5
Genomic Map