Assay Details
| Gene Symbol: IL17REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048934.1 | XP_017904423.1 | 9 - 10 | 3256 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL17REL| Gene Symbol: IL17REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048934.1 | XP_017904423.1 | 9 - 10 | 3256 | 79 | |
Set Membership: |
|