Assay Details
| Gene Symbol: IL17REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048934.1 | XP_017904423.1 | 1 - 2 | 2622 | 108 | |
| GenBank mRNA | JO614510.2 | - | 2 - 3 | 178 | 108 | |
More Information
Set Membership: |
|

IL17REL
Genomic Map