Assay Details
| Gene Symbol: LOC108636441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO427716.1 | - | 14 - 15 | 2045 | 91 | |
| Gene Symbol: LOC102170412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051138.1 | XP_017906627.1 | 14 - 15 | 2007 | 91 | |
| XM_018051139.1 | XP_017906628.1 | 13 - 14 | 2037 | 91 | ||
| GenBank mRNA | JO576964.1 | - | 24 - 25 | 3328 | 91 | |
More Information
Set Membership: |
|

LOC108636441
Genomic Map