Assay Details
| Gene Symbol: LOC108636441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051676.1 | XP_017907165.1 | 1 - 2 | 274 | 75 | |
| Gene Symbol: LOC102170412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO576964.1 | - | 4 - 5 | 753 | 75 | |
More Information
Set Membership: |
|

LOC108636441
Genomic Map