Assay Details
| Gene Symbol: LOC102175120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965609.2 | XP_013821063.1 | 22 - 23 | 4241 | 87 | |
| XM_018050275.1 | XP_017905764.1 | 23 - 24 | 4299 | 87 | ||
| XM_018050276.1 | XP_017905765.1 | 23 - 24 | 4299 | 87 | ||
| XM_018050277.1 | XP_017905766.1 | 22 - 23 | 4242 | 87 | ||
| GenBank mRNA | JO600138.2 | - | 12 - 13 | 3690 | 87 | |
More Information
Set Membership: |
|

LOC102175120
Genomic Map