Assay Details
| Gene Symbol: LOC102175120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965609.2 | XP_013821063.1 | 29 - 30 | 6998 | 86 | |
| XM_018050275.1 | XP_017905764.1 | 30 - 31 | 7056 | 86 | ||
| XM_018050276.1 | XP_017905765.1 | 30 - 31 | 7056 | 86 | ||
| XM_018050277.1 | XP_017905766.1 | 29 - 30 | 6999 | 86 | ||
| GenBank mRNA | JO600138.2 | - | 5 - 6 | 933 | 86 | |
More Information
Set Membership: |
|

LOC102175120
Genomic Map