Assay Details
| Gene Symbol: LOC102175120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965609.2 | XP_013821063.1 | 30 - 31 | 7174 | 95 | |
| XM_018050276.1 | XP_017905765.1 | 31 - 32 | 7232 | 95 | ||
More Information
Set Membership: |
|

LOC102175120
Genomic Map