Assay Details
| Gene Symbol: CTCFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968705.2 | XP_013824159.2 | 6 - 7 | 1355 | 63 | |
| XM_013968706.2 | XP_013824160.2 | 7 - 8 | 1274 | 63 | ||
More Information
Set Membership: |
|

CTCFL
Genomic Map