Assay Details
| Gene Symbol: C13H20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968817.1 | XP_013824271.1 | 2 - 3 | 813 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C13H20orf173| Gene Symbol: C13H20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968817.1 | XP_013824271.1 | 2 - 3 | 813 | 78 | |
Set Membership: |
|