Assay Details
| Gene Symbol: TOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057894.1 | XP_017913383.1 | 3 - 4 | 443 | 60 | |
| XM_018057895.1 | XP_017913384.1 | 3 - 4 | 443 | 60 | ||
| XM_018057896.1 | XP_017913385.1 | 2 - 3 | 362 | 60 | ||
| GenBank mRNA | JO647706.2 | - | 3 - 4 | 428 | 60 | |
More Information
Set Membership: |
|

TOX2
Genomic Map