Assay Details
| Gene Symbol: LOC102181889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060895.1 | XP_017916384.1 | 2 - 3 | 528 | 86 | |
| GenBank mRNA | JO439232.2 | - | 1 - 2 | 86 | ||
More Information
Set Membership: |
|

LOC102181889
Genomic Map