Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972362.2 | XP_013827816.1 | 4 - 5 | 637 | 56 | |
| GenBank mRNA | KA330158.1 | - | 2 - 3 | 215 | 56 | |
More Information
Set Membership: |
|

RHCE
Genomic Map