Assay Details
| Gene Symbol: LOC102180829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043118.1 | XP_017898607.1 | 9 - 10 | 1077 | 74 | |
| XM_018043119.1 | XP_017898608.1 | 9 - 10 | 1077 | 74 | ||
| XM_018043120.1 | XP_017898609.1 | 9 - 10 | 1077 | 74 | ||
| XM_018043123.1 | XP_017898612.1 | 8 - 9 | 892 | 74 | ||
| XM_018043124.1 | XP_017898613.1 | 8 - 9 | 783 | 74 | ||
| XM_018043125.1 | XP_017898614.1 | 8 - 9 | 887 | 74 | ||
| XM_018043126.1 | XP_017898615.1 | 2 - 3 | 203 | 74 | ||
More Information
Set Membership: |
|

LOC102180829
Genomic Map