Assay Details
| Gene Symbol: LOC102180829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043118.1 | XP_017898607.1 | 2 - 3 | 266 | 77 | |
| XM_018043119.1 | XP_017898608.1 | 2 - 3 | 266 | 77 | ||
| XM_018043120.1 | XP_017898609.1 | 2 - 3 | 266 | 77 | ||
| XM_018043121.1 | XP_017898610.1 | 1 - 2 | 61 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018043121.1 transcripts.
Set Membership: |
|

LOC102180829
Genomic Map