Assay Details
| Gene Symbol: LOC102180829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043118.1 | XP_017898607.1 | 3 - 4 | 374 | 106 | |
| XM_018043119.1 | XP_017898608.1 | 3 - 4 | 374 | 106 | ||
| XM_018043120.1 | XP_017898609.1 | 3 - 4 | 374 | 106 | ||
| XM_018043121.1 | XP_017898610.1 | 2 - 3 | 169 | 106 | ||
| GenBank mRNA | KA306997.1 | - | 6 - 7 | 578 | 106 | |
More Information
Set Membership: |
|

LOC102180829
Genomic Map