Assay Details
| Gene Symbol: LOC102180829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043118.1 | XP_017898607.1 | 4 - 5 | 457 | 119 | |
| XM_018043119.1 | XP_017898608.1 | 4 - 5 | 457 | 119 | ||
| XM_018043120.1 | XP_017898609.1 | 4 - 5 | 457 | 119 | ||
| XM_018043121.1 | XP_017898610.1 | 3 - 4 | 252 | 119 | ||
| XM_018043123.1 | XP_017898612.1 | 3 - 4 | 272 | 119 | ||
| XM_018043124.1 | XP_017898613.1 | 3 - 4 | 163 | 119 | ||
| XM_018043125.1 | XP_017898614.1 | 3 - 4 | 267 | 119 | ||
| XM_018043127.1 | XP_017898616.1 | 1 - 2 | 88 | 119 | ||
| GenBank mRNA | KA306997.1 | - | 5 - 6 | 495 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018043127.1 transcripts.
Set Membership: |
|

LOC102180829
Genomic Map