Assay Details
| Gene Symbol: LOC102180829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043118.1 | XP_017898607.1 | 5 - 6 | 513 | 70 | |
| XM_018043119.1 | XP_017898608.1 | 5 - 6 | 513 | 70 | ||
| XM_018043120.1 | XP_017898609.1 | 5 - 6 | 513 | 70 | ||
| XM_018043121.1 | XP_017898610.1 | 4 - 5 | 308 | 70 | ||
| XM_018043123.1 | XP_017898612.1 | 4 - 5 | 328 | 70 | ||
| XM_018043124.1 | XP_017898613.1 | 4 - 5 | 219 | 70 | ||
| XM_018043125.1 | XP_017898614.1 | 4 - 5 | 323 | 70 | ||
| GenBank mRNA | KA306997.1 | - | 4 - 5 | 439 | 70 | |
More Information
Set Membership: |
|

LOC102180829
Genomic Map