Assay Details
Gene Symbol: LOC102180829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043118.1 | XP_017898607.1 | 6 - 7 | 586 | 68 | |
XM_018043119.1 | XP_017898608.1 | 6 - 7 | 586 | 68 | ||
XM_018043120.1 | XP_017898609.1 | 6 - 7 | 586 | 68 | ||
XM_018043121.1 | XP_017898610.1 | 5 - 6 | 381 | 68 | ||
XM_018043123.1 | XP_017898612.1 | 5 - 6 | 401 | 68 | ||
XM_018043124.1 | XP_017898613.1 | 5 - 6 | 292 | 68 | ||
XM_018043125.1 | XP_017898614.1 | 5 - 6 | 396 | 68 | ||
GenBank mRNA | KA306997.1 | - | 3 - 4 | 366 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
