Assay Details
| Gene Symbol: LOC102180829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043118.1 | XP_017898607.1 | 7 - 8 | 777 | 92 | |
| XM_018043119.1 | XP_017898608.1 | 7 - 8 | 777 | 92 | ||
| XM_018043120.1 | XP_017898609.1 | 7 - 8 | 777 | 92 | ||
| XM_018043121.1 | XP_017898610.1 | 6 - 7 | 572 | 92 | ||
| XM_018043123.1 | XP_017898612.1 | 6 - 7 | 592 | 92 | ||
| XM_018043124.1 | XP_017898613.1 | 6 - 7 | 483 | 92 | ||
| XM_018043125.1 | XP_017898614.1 | 6 - 7 | 587 | 92 | ||
| GenBank mRNA | KA306997.1 | - | 2 - 3 | 175 | 92 | |
More Information
Set Membership: |
|

LOC102180829
Genomic Map