Assay Details
| Gene Symbol: PID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314346.1 | NP_001301275.1 | 2 - 3 | 195 | 95 | |
| GenBank mRNA | JN257257.1 | - | 2 - 3 | 195 | 95 | |
| JO554384.2 | - | 2 - 3 | 315 | 95 | ||
| KT984176.1 | - | 2 - 3 | 188 | 95 | ||
More Information
Set Membership: |
|

PID1
Genomic Map