Assay Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680867.2 | XP_005680924.1 | 2 - 3 | 284 | 87 | |
| GenBank mRNA | JT689609.1 | - | 3 - 4 | 431 | 87 | |
More Information
Set Membership: |
|

KLRF1
Genomic Map