Assay Details
| Gene Symbol: CCN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053051.1 | XP_017908540.1 | 4 - 5 | 853 | 71 | |
| GenBank mRNA | JR114848.1 | - | 2 - 3 | 239 | 71 | |
| JT632286.1 | - | 1 - 2 | 269 | 71 | ||
More Information
Set Membership: |
|

CCN6
Genomic Map