Assay Details
| Gene Symbol: MTFMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685304.3 | XP_005685361.3 | 5 - 6 | 791 | 79 | |
| XM_018053660.1 | XP_017909149.1 | 4 - 5 | 581 | 79 | ||
More Information
Set Membership: |
|

MTFMT
Genomic Map