Assay Details
| Gene Symbol: NCR3LG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689580.3 | XP_005689637.2 | 3 - 4 | 978 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NCR3LG1| Gene Symbol: NCR3LG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689580.3 | XP_005689637.2 | 3 - 4 | 978 | 86 | |
Set Membership: |
|