Assay Details
| Gene Symbol: TMEM144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691302.2 | XP_005691359.1 | 6 - 7 | 739 | 72 | |
| XM_018061483.1 | XP_017916972.1 | 6 - 7 | 739 | 72 | ||
| GenBank mRNA | JO625930.2 | - | 6 - 7 | 811 | 72 | |
More Information
Set Membership: |
|

TMEM144
Genomic Map