Assay Details
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691566.3 | XP_005691623.2 | 1 - 2 | 697 | 98 | |
| GenBank mRNA | KA321907.1 | - | 3 - 4 | 1065 | 98 | |
More Information
Set Membership: |
|

GATC
Genomic Map