Assay Details
| Gene Symbol: COTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691841.3 | XP_005691898.1 | 2 - 3 | 387 | 62 | |
| GenBank mRNA | JO498636.2 | - | 2 - 3 | 383 | 62 | |
More Information
Set Membership: |
|

COTL1
Genomic Map