Assay Details
| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695485.3 | XP_005695542.1 | 6 - 7 | 516 | 73 | |
| XM_018038281.1 | XP_017893770.1 | 6 - 7 | 516 | 73 | ||
| XM_018038282.1 | XP_017893771.1 | 6 - 7 | 516 | 73 | ||
| XM_018038283.1 | XP_017893772.1 | 6 - 7 | 516 | 73 | ||
| XM_018038284.1 | XP_017893773.1 | 5 - 6 | 440 | 73 | ||
| XM_018038285.1 | XP_017893774.1 | 6 - 7 | 477 | 73 | ||
| GenBank mRNA | JO605864.2 | - | 3 - 4 | 300 | 73 | |
More Information
Set Membership: |
|

UBE2D4
Genomic Map