Assay Details
| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038281.1 | XP_017893770.1 | 7 - 8 | 551 | 88 | |
| XM_018038282.1 | XP_017893771.1 | 7 - 8 | 551 | 100 | ||
| XM_018038284.1 | XP_017893773.1 | 6 - 7 | 475 | 88 | ||
| XM_018038285.1 | XP_017893774.1 | 7 - 8 | 512 | 88 | ||
| GenBank mRNA | JO605864.2 | - | 2 - 3 | 265 | 88 | |
More Information
Set Membership: |
|

UBE2D4
Genomic Map