Assay Details
| Gene Symbol: NMRAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040384.1 | XP_017895873.1 | 2 - 3 | 250 | 55 | |
| XM_018040385.1 | XP_017895874.1 | 2 - 3 | 172 | 55 | ||
| XM_018040386.1 | XP_017895875.1 | 2 - 3 | 98 | 55 | ||
| GenBank mRNA | KA323729.1 | - | 4 - 5 | 981 | 55 | |
More Information
Set Membership: |
|

NMRAL1
Genomic Map