Assay Details
| Gene Symbol: LOC102190194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040746.1 | XP_017896235.1 | 11 - 12 | 1346 | 97 | |
| XM_018040748.1 | XP_017896237.1 | 10 - 11 | 1268 | 97 | ||
| XM_018040749.1 | XP_017896238.1 | 9 - 10 | 1108 | 97 | ||
More Information
Set Membership: |
|

LOC102190194
Genomic Map